Learn More
Cayman Chemical Fulacimstat, Purity: ≥98%, CAS: 1488354-15-9, 5 mg
SDP
Catalog No. 502916502
Encompass_Preferred
A chymase inhibitor (IC50s = 4 and 3 nM for the human and hamster enzyme, respectively); reduces isoprenaline-induced cardiac fibrosis in hamsters at 1, 3, and 10 mg/kg; inhibits decreases in left ventricular ejection fraction and increases in left ventricular end-systolic volume, decreases in plasma levels of nt-proBNP, TGF-β1, and TNF-α, as well as the levels of interstitial fibrosis and the cross-sectional area of myocytes, in a coronary microembolism-induced canine model of heart failure at 2 and 5 mg/kg per day
Catalog No. 50-291-6502 Supplier Cayman Chemical Supplier No. 377315 MG
A chymase inhibitor (IC50s = 4 and 3 nM for the human and hamster enzyme, respectively); reduces isoprenaline-induced cardiac fibrosis in hamsters at 1, 3, and 10 mg/kg; inhibits decreases in left ventricular ejection fraction and increases in left ventricular end-systolic volume, decreases in plasma levels of nt-proBNP, TGF-β1, and TNF-α, as well as the levels of interstitial fibrosis and the cross-sectional area of myocytes, in a coronary microembolism-induced canine model of heart failure at 2 and 5 mg/kg per day