Learn More
Sigma Aldrich Fine Chemicals Biosciences Folin & Ciocalteu's phenol reagent suitable for determination of total protein by Lowry method, 2 N | MFCD00132625 | 1L
Catalog No. NC1865725
Encompass
Folin & Ciocalteu's phenol reagent suitable for determination of total protein by Lowry method, 2 N | MFCD00132625 | 1L
Catalog No. NC1865725 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F92521L
Folin & Ciocalteu's phenol reagent suitable for determination of total protein by Lowry method, 2 N | MFCD00132625 | 1L