Learn More
Medicus Health Full Tube Racks - Polypropylene for 16mm Tubes, Blue
SDP
Catalog No. NC3304146
Encompass
Full Tube Racks, Polypropylene for 16mm Tubes, Blue are molded in a single, continuous piece no assembly required and no detachable pieces
∙Test Tube Racks have 72 places for up to 16mm diameter tubes
∙ Racks offer good chemical and heat resistance (0-110° C)
∙ Include molded-in index coordinates and flat areas for labeling
∙ Mix and match any color and quantity
∙ Available in four colors Red, White, Blue and Green, please specify color when ordering
∙ Will not float
∙ Dimensions: 9.8"L x 5"W x 2..75"H
Catalog No. NC3304146 Supplier Medicus Health Supplier No. 2286M
Full Tube Racks, Polypropylene for 16mm Tubes, Blue are molded in a single, continuous piece no assembly required and no detachable pieces
∙Test Tube Racks have 72 places for up to 16mm diameter tubes
∙ Racks offer good chemical and heat resistance (0-110° C)
∙ Include molded-in index coordinates and flat areas for labeling
∙ Mix and match any color and quantity
∙ Available in four colors Red, White, Blue and Green, please specify color when ordering
∙ Will not float
∙ Dimensions: 9.8"L x 5"W x 2..75"H