Learn More
Medicus Health Full 16mm Tube Racks - Ultra Durable Delrin, Red
SDP
Catalog No. NC2372230
Encompass
Full Tube Racks - Ultra Durable Delrin for 16mm Tubes Holds up to seventy two tubes 16mm in diameter. Tube Rack made from Delrin designed for the toughest conditions. Constructed of Delrin material so they can handle extreme cold and hot temperatures (-40 F - 250 F). Resistant to acids, solvents, and most chemicals. Has 72 places for up to 16mm diameter tubes. Available in four colors Red, White, Blue and Green - Please specify color when ordering. Mix and match any color and quantity. Includes molded-in index coordinates and flat areas for labeling. Autoclavable. Dimensions: 9.8""""L x 5""""W x 2.75""""H
Catalog No. NC2372230 Supplier Medicus Health Supplier No. 5259M1RED
Full Tube Racks - Ultra Durable Delrin for 16mm Tubes Holds up to seventy two tubes 16mm in diameter. Tube Rack made from Delrin designed for the toughest conditions. Constructed of Delrin material so they can handle extreme cold and hot temperatures (-40 F - 250 F). Resistant to acids, solvents, and most chemicals. Has 72 places for up to 16mm diameter tubes. Available in four colors Red, White, Blue and Green - Please specify color when ordering. Mix and match any color and quantity. Includes molded-in index coordinates and flat areas for labeling. Autoclavable. Dimensions: 9.8""""L x 5""""W x 2.75""""H