Learn More
Hach Company Full Container Shut-Off for Refrigerated and All-Weather Samplers
Catalog No. NC3036732 Shop All Hach Company Products
Encompass
Full container shut-off accessory for refrigerated and all-weather samplers. It prevents overfilling by automatically stopping sample collection when the collection container reaches capacity, reducing spills and protecting sampler components during automated composite sampling operations.
  • Prevents overfilling by stopping sample collection when bottle is full.
  • Compatible with refrigerated and all-weather samplers.
  • Helps reduce spills and sample loss during automated sampling.
  • Simple installation and integration with existing sampler setups.
  • Ideal for composite and automated field sampling applications.
Catalog No. NC3036732 Supplier Hach Company Supplier No. 8847
Full container shut-off accessory for refrigerated and all-weather samplers. It prevents overfilling by automatically stopping sample collection when the collection container reaches capacity, reducing spills and protecting sampler components during automated composite sampling operations.
  • Prevents overfilling by stopping sample collection when bottle is full.
  • Compatible with refrigerated and all-weather samplers.
  • Helps reduce spills and sample loss during automated sampling.
  • Simple installation and integration with existing sampler setups.
  • Ideal for composite and automated field sampling applications.