Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Full Coverage Face Shields with Anti-Fog

Catalog No. 03411844 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Lens Coating:
Anti-fog
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
03-411-844 Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 03-411-844 Supplier Bel-Art™ Supplier No. H248741000
  • $380.00
    $314.65 / Case of 20

    Save $65.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protects against exposure to bodily fluids and infectious materials

  • Large, wrap-around design provides coverage of entire face with unrestricted visibility
  • Helps protect the face from potential hazards such as dust, dirt, pathogens, sprays and fluids
  • Anti-fog coating on interior surface prevents fogging for clear vision
  • Comfortable; suited for extended wear
  • Thick top foam guard provides distance between user and shield
  • Clear PET plastic viewing area; remove protective film prior to use
  • Disposable shield; single use only
  • Notes:
    • Full Coverage Face Shields with Anti-Fog have not been FDA cleared or approved
    • The product has been authorized by FDA under an EUA for use by healthcare providers as personal protective equipment.
    • This product is only authorized for the duration of the declaration that circumstances justifying the authorization of emergency use under Section 564(b)(1) of the Act, 21 USC 360bbb-3(b)(1) unless the authorization is terminated or revoked sooner.
  • Specifications

    Width (English) 13 in.
    Certifications/Compliance CE
    Height (English) 9 in.
    Height (Metric) 23 cm
    Width (Metric) 33 cm
    Thickness (Metric) 0.36 mm
    Lens Coating Anti-fog
    Lens Tint None
    Product Type Full Coverage Faceshield
    Quantity 20/Pk.
    Material PET
    Disposable Single-use
    Lens Material PET
    Frame Material Plastic
    UV Protection None
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.