Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CleanSpace™ Full Face PAPR Mask

Catalog No. 17800610 Shop All Cleanspace Technology Products
Change view
Click to view available options
Size:
Medium/Large
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-800-610 Small
17-800-609 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-800-610 Supplier CleanSpace™ Supplier No. PAF1106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Full face mask is suitable for heavy, long term work, or exposure. Designed for use with the CleanSpace™ Ultra and EX PAPR systems.

  • Used with the revolutionary CleanSpace™PAPR: lightweight, no hoses or belts
  • Wide panoramic visor
  • Hard scratch and solvent resistant coating
  • Suitable for long periods of wear
  • Positive pressure airflow and inner Orinasal mask prevents fogging
  • Easy breathing and no resistance even during heavy exertion
  • Visor seal silicone design seals to the face for optimum protection and comfort.
  • Inner Orinasal mask with unique clip-in clip-out design is easily removed for cleaning
  • Mylar speech diaphragm for clear communication on the job
  • Age resistant materials
  • PortaCount testing enabled for quantitative mask fit testing (Sold separately)
  • Spectacle kit: Fully adjustable for all face sizes, wide frames to fit large lenses for peripheral vision; engineered stability during vigorous activity (Sold separately)
  • Tear-Offs: Visor protection against scratches (Sold separately)
  • Hard moulded polycarbonate visor; Silicone face seal and Inner Orinasal Mask cushion. Nylon mask clips; Mylar speech diaphragm. EPDM rubber harness; impact modified reinforced polypropylene exhalation valve housing; silicone O-rings

Specifications

Certifications/Compliance AS/NZS1716: 2012
Certifications/Compliance (Europe) EN 12942
Product Type PAPR Full Face Mask
Size Small
Facepiece/Headtop Model PAPR full face mask only
For Use With (Equipment) PAF-1070
Model PAF-1106
Storage Requirements Store at -10°C to 55°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.