Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CleanSpace™ Technology Full Face Mask Tear-Off Visor Anti-Scratch Protectors

Catalog No. 17800625 Shop All Cleanspace Technology Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-800-625 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-800-625 Supplier CleanSpace™ Technology Supplier No. PAF1018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Full face mask tear-off visor anti-scratch protectors are protective films for the full face mask visor.

  • Pack of 10 tear offs
  • Clear and easy to use
  • Used with the revolutionary CleanSpace™ Respirator: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement

Specifications

Certifications/Compliance AS/NZS1716: 2012, EN 12942
Disposable Single-use
Material Polyester
Product Type Tear off Visor Anti-Scratch Protectors
For Use With (Equipment) PAF-1014, PAF-1106
Model PAF-1018
Quantity 10/Pk.
Storage Requirements Store at -10°C to +55°C
Dimensions (L x W x H) 210 x 125 mm
Weight (Metric) 75 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.