Learn More
Dentec Safety™ Full Face Respirator Replacement Lens
SDP
Catalog No. 19222481 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Polycarbonate
Catalog No.
19-222-481 Polycarbonate
1 options
Catalog No. 19-222-481 Supplier Dentec Safety™ Supplier No. 1307086
Add to Cart
Add to Cart
Replacement lens for 130M Series full face respirator.
  • Accessory
Specifications
Disposable Reusable
Material Polycarbonate
Product Type Facepiece Lens
For Use With (Equipment) 130M Series full face respirator
Includes Lens, Cover
Model 1307086