Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Full-Finger Medical Nylon Glove Liners
GSA_VA

Catalog No. 19044150 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-044-150 Small
19-044-151 Medium
19-044-152 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-044-150 Supplier Wells Lamont™ Supplier No. M115S
  • $116.00
    $91.65 / Pack of 25

    Save $24.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $401.00
    $319.65 / Case of 4 PK

    Save $81.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Full-Finger Medical Nylon Glove Liners are constructed of high-quality, 100% continuous filament nylon and packaged in a secure box. Wear under a disposable glove for additional comfort, or to reduce hand fatigue from perspiration.

  • 100% continuous nylon filament creates an ideal barrier between hands and disposable gloves
  • Reduces allergies and adverse reactions that can be caused by prolonged exposure to latex
  • Packaged in a secure box so that liners remain clean
  • Fully launderable up to 20 times without shrinking
  • Full finger, regular cuff design
  • Non-sterile
  • Made in the USA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Applications Healthcare
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Nylon Glove Liner
Size Small
Sterility Non-sterile
Finger Style Full
For Use With (Application) Reducing Latex Allerigies and Adverse Reactions
Glove Cuff Style Knit
Hem Color Green
Packaging Type Box
Series M115
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.