Learn More
Quality Lab Accessories 70 Micron Porous Filters, 100 pack
Supplier:  Quality Lab Accessories FIL07001A
Encompass
70 Micron Porous Filters, UHMW Polyethylene, Agilent/VanKel/Varian compatible (Pack/100)
Catalog No. NC9041112