Learn More
FLUKE ELECTRONICS CORPORATION FULL IMMERSION PRT PROBE 50MM
Catalog No. NC2602508
Encompass
5606 full Immersion Probe, 50mm - 200 to 160C for 1523/1524
Catalog No. NC2602508 Supplier FLUKE ELECTRONICS CORPORATION Supplier No. 560650P
5606 full Immersion Probe, 50mm - 200 to 160C for 1523/1524