Learn More
World Precision Instrument Full Micro Perforated Trays with Lids
SDP
Supplier:  World Precision Instrument WP2355
Encompass
  • Manufactured of stainless steel grade AISI-304
  • Electropolished
  • Full wall perforation to protect sharp instruments
  • Protects edges and tips from damage
  • Smooth rounded corners
  • Perforation hole size is 2.0 mm
  • Hinged lids
Catalog No. 50-209-0155