Learn More
RNA Medical FULL RANGE CO-OX CTRL
SDP
Catalog No. NC9980296
Encompass
QC 253 is an assayed quality control material used for monitoring the measurements of total hemoglobin and hemoglobin fractions on CO-Oximeter instrumentation. It provides an alternative for those needing a control that parallels the human physiologic range for tHb, O2Hb, and COHb. •Requires refrigeration storage •18 month shelf life •Available in single or multi-level boxes •Peer comparison with optional on-line data submission Analytes tHb, O2Hb, COHb, MetHb, VolO2(%) (O2Ct), RHb (HHb)
Catalog No. NC9980296 Supplier RNA Medical Supplier No. QC 253
 Restricted Item
Add to Cart
Add to Cart
QC 253 is an assayed quality control material used for monitoring the measurements of total hemoglobin and hemoglobin fractions on CO-Oximeter instrumentation. It provides an alternative for those needing a control that parallels the human physiologic range for tHb, O2Hb, and COHb. •Requires refrigeration storage •18 month shelf life •Available in single or multi-level boxes •Peer comparison with optional on-line data submission Analytes tHb, O2Hb, COHb, MetHb, VolO2(%) (O2Ct), RHb (HHb)