Learn More
Beckman Coulter Full Reservoir Sterile, hold up to 150 mL
Catalog No. NC9209479
Encompass
Full reservoir (150mL) The Modular Reservoir System holds liquids for pipetting with Biomek Pipette Tools and Beckman Coulter Manual Pipettors
Catalog No. NC9209479 Supplier Beckman Coulter Supplier No. 372784
Full reservoir (150mL) The Modular Reservoir System holds liquids for pipetting with Biomek Pipette Tools and Beckman Coulter Manual Pipettors