Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Science Lab Activities Complete Set of 12
GREENER_CHOICESCI_ED

Catalog No. s05884 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05884 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, Lead and Lead Compounds, Phenolphthalein, Methanol/ Methyl Isobutyl Ketone, Respirable Crystalline Silica, Acetyl Salicylic Acid/ Aspirin and, Carbon Black, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S05884 Supplier Innovating Science™ Supplier No. IS9000
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, Lead and Lead Compounds, Phenolphthalein, Methanol/ Methyl Isobutyl Ketone, Respirable Crystalline Silica, Acetyl Salicylic Acid/ Aspirin and, Carbon Black, which are known to the State of California to cause cancer and reproductive harm.

Includes

Complete set includes these popular activity kits:

  • Gunshot Residue Presumptive Test Kit (S96637)
  • Presumptive Blood Test Kit (S96638)
  • Forensic Chemistry of Document Analysis Kit (S96639)
  • Physical Properties of Glass Kit (S96641)
  • Forensic Chemistry of Unknown Substances Kit (S96642)
  • Forensic Chemistry of Hair Analysis Kit (S96643)
  • Forensic Chemistry of Blood Types Kit (S96644)
  • Chemiluminescence in Blood Stain Detection (S96645)
  • Forensic Chemistry of Drug Detection Kit (S96646)
  • Crime Scene Investigation Lab (S96647)
  • Forensic Chemistry of Dusting for Fingerprints Set (S96648)
  • Forensic Chemistry: Chemical Detection of Fingerprints (S96357)

Enjoy the convenience of having the series of forensic test kits available in your classroom.

Introduce tudents to the investigation with several different types of possible evidence, such as; fingerprints, blood detection/blood typing, identification of unknown substances, gunshot residue, properties of glass, and more

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) For forensic test
Includes Gunshot Residue Presumptive Test Kit (S96637), Presumptive Blood Test Kit (S96638), Forensic Chemistry of Document Analysis Kit (S96639), Physical Properties of Glass Kit (S96641), Forensic Chemistry of Unknown Substances Kit (S96642), Forensic Chemistry of Hair Analysis Kit (S96643), Forensic Chemistry of Blood Types Kit (S96644), Chemiluminescence in Blood Stain Detection (S96645), Forensic Chemistry of Drug Detection Kit (S96646), Crime Scene Investigation Lab (S96647), Forensic Chemistry of Dusting for Fingerprints Set (S96648), Forensic Chemistry: Chemical Detection of Fingerprints (S96357)
Quantity 1
Dimensions (L x W x H) 21 x 16 x 17 in.
Class Size 15 Lab Groups
Product Type Curriculum Forensic Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.