Learn More
ILC Dover Full Skirt Hood for Sentinel XT Blower
Catalog No. 19631078
Change view
Click to view available options
Quantity:
10/Pk.
25/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-631-078 25/Pk.
19-631-077 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-631-078 Supplier ILC Dover Supplier No. S311325
Add to Cart
Add to Cart
Full skirt hood has an APF of 1,000 and can be used with the Sentinel XL and XT Blower Systems.
  • Ideal for both healthcare and pharmaceutical industries
Specifications
Assigned Protection Factor (APF) 1000
Product Type Full Skirt Hood
For Use With (Application) Healthcare, Pharmaceutical
For Use With (Equipment) Sentinel XT Blower
Model S-3113-25
Quantity 25/Pk.