Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILC Dover Sentinel XT Non-Incendive Blower (Blue)

Catalog No. 19631079
Change view
Click to view available options
Color: Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-631-079 Color: Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-631-079 Supplier ILC Dover Supplier No. S3315
Only null left
Add to Cart
Add to Cart

Sentinel XT Non-Incendive Blower (Blue) delivers 8+ CFM airflow, exceeding NIOSH requirements.

  • Approved for use in hazardous locations
  • Secure lock prevents battery removal without use of a tool
  • Brushless motor design provides long life and quiet operation
  • Secondary impact protection chamber for air handling unit
  • Approved by Factory Mutual for use in Class I, II, and III; Division 2; Groups C, D, E, F and G

Specifications

Certifications/Compliance Factory Mutual for use in Class I, II, and III; Division 2; Groups C, D, E, F and G
Color Blue
Product Type Motor Blower
Flow Rate 8.0 CFM
For Use With (Application) Pharmaceutical
For Use With (Equipment) Sentinel XT Systems
Model S-3315
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.