Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fuller's Earth (Powder/Technical), Fisher Chemical

Catalog No. F903 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
3 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
F90-3 3 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F90-3

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8031-18-3
MDL Number MFCD00081617

Specifications

Color Yellow
Physical Form Powder/Solid
Quantity 3 kg
Grade Technical
Identification Pass Test
Packaging Poly Pail
Chemical Name or Material Fuller's Earth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.