Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fullerene-C60 | 99685-96-8 | MFCD00151408 | 99.7% | 720.64 g/mol | C60 | 25 MG
SDP

Catalog No. 5000303959
Encompass_Preferred

Fullerene-C60 is the spherical carbon allotrope C60 supplied as a brown-to-black solid reagent for materials science and biochemical research. The product is characterized by high reported purity and defined storage recommendations to preserve stability; common applications include nanomaterials studies, electron-transfer experiments, and biochemical assays.

  • High reported purity: 99.73%.
  • Solid, brown to black powder form for convenient handling.
  • Molecular weight 720.64 g/mol; formula C60.
  • Available in small-mass laboratory quantities for research use.
  • Recommended storage: powder at -20°C for long term, 4°C for shorter term.
  • Suitable for nanomaterials research, physical chemistry, and biochemical assays.

Catalog No. 50-003-03959 Supplier Medchemexpress LLC Supplier No. 25MGHYW020768
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fullerene-C60 is the spherical carbon allotrope C60 supplied as a brown-to-black solid reagent for materials science and biochemical research. The product is characterized by high reported purity and defined storage recommendations to preserve stability; common applications include nanomaterials studies, electron-transfer experiments, and biochemical assays.

  • High reported purity: 99.73%.
  • Solid, brown to black powder form for convenient handling.
  • Molecular weight 720.64 g/mol; formula C60.
  • Available in small-mass laboratory quantities for research use.
  • Recommended storage: powder at -20°C for long term, 4°C for shorter term.
  • Suitable for nanomaterials research, physical chemistry, and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.