Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fullerene-C60 | 99685-96-8 | MFCD00151408 | 99.7% | 720.64 g/mol | C60 | 50 MG
SDP

Catalog No. 5000299572
Encompass_Preferred

Fullerene-C60 is a buckminsterfullerene (C60) molecular carbon compound supplied as a brown to black solid for biochemical and analytical research. It has molecular weight 720.64 g/mol and CAS 99685-96-8, and is provided at high purity for experimental use.

  • High purity: 99.73%.
  • Molecular formula: C60; molecular weight 720.64 g/mol.
  • Physical form: brown to black solid powder.
  • Low solubility: H2O < 0.1 mg/mL; DMSO < 1 mg/mL.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years).
  • Suitable for biochemical assays and analytical studies.

Catalog No. 50-002-99572 Supplier Medchemexpress LLC Supplier No. HYW02076850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fullerene-C60 is a buckminsterfullerene (C60) molecular carbon compound supplied as a brown to black solid for biochemical and analytical research. It has molecular weight 720.64 g/mol and CAS 99685-96-8, and is provided at high purity for experimental use.

  • High purity: 99.73%.
  • Molecular formula: C60; molecular weight 720.64 g/mol.
  • Physical form: brown to black solid powder.
  • Low solubility: H2O < 0.1 mg/mL; DMSO < 1 mg/mL.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years).
  • Suitable for biochemical assays and analytical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.