Learn More
Medchemexpress LLC Fullerene-C60-Ih (9CI, ACI) | 99685-96-8 | MFCD00151408 | 99.7% | 720.64 g·mol⁻¹ | C60 | 100 MG
SDP
Catalog No. 5000301322
Encompass_Preferred
Fullerene-C60 (C60) is a high-purity carbon nanomaterial (buckminsterfullerene) used in photophysical and biochemical research. It serves as an effective electron acceptor and radical scavenger, and efficiently produces singlet oxygen upon light excitation, making it useful for photodynamic studies and electron-transfer experiments.
  • High purity suitable for laboratory research.
  • Efficient singlet oxygen generation under UV/visible light.
  • Strong electron-accepting and radical-scavenging properties.
  • Available in small pack sizes for experimental flexibility.
  • Well-characterized molecular weight and composition.
Catalog No. 50-003-01322 Supplier Medchemexpress LLC Supplier No. HYW020768100MG
Fullerene-C60 (C60) is a high-purity carbon nanomaterial (buckminsterfullerene) used in photophysical and biochemical research. It serves as an effective electron acceptor and radical scavenger, and efficiently produces singlet oxygen upon light excitation, making it useful for photodynamic studies and electron-transfer experiments.
  • High purity suitable for laboratory research.
  • Efficient singlet oxygen generation under UV/visible light.
  • Strong electron-accepting and radical-scavenging properties.
  • Available in small pack sizes for experimental flexibility.
  • Well-characterized molecular weight and composition.