Learn More
Strem, An Ascensus Company CAS# 131159-39-2. 5g. Fullerene carbon soot (contains 5-8wt% C60/C70 and higher fullerenes). MFCD00282904
SDP
Catalog No. 503657
Encompass
CAS# 131159-39-2. 5g. Fullerene carbon soot (contains 5-8wt% C60/C70 and higher fullerenes). MFCD00282904. Molecular Weight: . Molecular Formula: C60/C70. Color/form: black pwdr. Strem# 06-0512. http://www.strem.com/catalog/v/06-0512/
Catalog No. 50-365-7 Supplier Strem, An Ascensus Company Supplier No. 0605125G
CAS# 131159-39-2. 5g. Fullerene carbon soot (contains 5-8wt% C60/C70 and higher fullerenes). MFCD00282904. Molecular Weight: . Molecular Formula: C60/C70. Color/form: black pwdr. Strem# 06-0512. http://www.strem.com/catalog/v/06-0512/