Learn More
Medchemexpress LLC HY-13636 100mg Medchemexpress, Fulvestrant CAS:129453-61-8 Purity:>98%
SDP
Catalog No. 502029016
Encompass_Preferred
Medchemexpress, HY-13636 100mg Fulvestrant CAS:129453-61-8 Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9016 Supplier Medchemexpress LLC Supplier No. HY13636100MG
Medchemexpress, HY-13636 100mg Fulvestrant CAS:129453-61-8 Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.