Learn More
Selleck Chemical LLC Fulvestrant 10mM/1mL 129453-61-8 ICI-182780, ZD 9238
SDP
Catalog No. 501361005
Encompass_Preferred
Fulvestrant (ICI-182780, ZD 9238) is an estrogen receptor (ER) antagonist with IC50 of 0.94 nM in a cell-free assay. Fulvestrant also induces autophagy and apoptosis and has antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1005 Supplier Selleck Chemical LLC Supplier No. S119110MM/1ML
Fulvestrant (ICI-182780, ZD 9238) is an estrogen receptor (ER) antagonist with IC50 of 0.94 nM in a cell-free assay. Fulvestrant also induces autophagy and apoptosis and has antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.