Learn More
Medchemexpress LLC Fulvestrant-3-boronic acid | 1853279-29-4 | 99.7% | 634.59 | 1 MG
SDP
Catalog No. 5000351745
Encompass_Preferred
Fulvestrant-3-boronic acid is an orally active ERα inhibitor that competitively binds to ERα (IC50 = 4.1 nM) and effectively degrades ERα in breast cancer cells.
  • Orally active ERα inhibitor
  • Competitively binds to ERα (IC50 = 4.1 nM)
  • Effectively degrades ERα in breast cancer cells
Catalog No. 50-003-51745 Supplier Medchemexpress LLC Supplier No. HY1371441MG
Fulvestrant-3-boronic acid is an orally active ERα inhibitor that competitively binds to ERα (IC50 = 4.1 nM) and effectively degrades ERα in breast cancer cells.
  • Orally active ERα inhibitor
  • Competitively binds to ERα (IC50 = 4.1 nM)
  • Effectively degrades ERα in breast cancer cells