Learn More
Medchemexpress LLC HY-13636 50mg , Fulvestrant CAS:129453-61-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1363650MG
Encompass_Preferred
HY-13636 50mg Fulvestrant CAS: 129453-61-8 Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant is also a GPR30 agonist. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9063