Learn More
Medchemexpress LLC Fulvestrant-d5 | 129453-61-8 | 99.66% | 611.80 | 1 MG
SDP
Catalog No. 5000349459
Encompass_Preferred
Fulvestrant-d5 (ICI 182780-d5) is an isotope of Fulvestrant and a potent estrogen receptor (ER) antagonist. It effectively inhibits the growth of ER-positive MCF-7 cells and induces autophagy, demonstrating antitumor efficacy. This compound serves as a valuable tool for various analytical applications.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes for quantitation during drug development
Catalog No. 50-003-49459 Supplier Medchemexpress LLC Supplier No. HY13636S11MG
Fulvestrant-d5 (ICI 182780-d5) is an isotope of Fulvestrant and a potent estrogen receptor (ER) antagonist. It effectively inhibits the growth of ER-positive MCF-7 cells and induces autophagy, demonstrating antitumor efficacy. This compound serves as a valuable tool for various analytical applications.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes for quantitation during drug development