Learn More
Medchemexpress LLC Fulvestrant (Standard) | 129453-61-8 | 99.9% | 606.77 | 1 MG
SDP
Catalog No. 5000350250
Encompass_Preferred
Fulvestrant (Standard) is the analytical standard of Fulvestrant, intended for research and analytical applications. It acts as a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant also functions as a GPR30 agonist, effectively inhibiting the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM, inducing autophagy, and demonstrating antitumor efficacy.
  • Analytical standard for research and analytical applications
  • Potent estrogen receptor (ER) antagonist
  • GPR30 agonist
  • Inhibits growth of ER-positive MCF-7 cells
  • Induces autophagy and demonstrates antitumor efficacy
  • Solid appearance
  • White to off-white color
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-003-50250 Supplier Medchemexpress LLC Supplier No. HY13636R1MG
Fulvestrant (Standard) is the analytical standard of Fulvestrant, intended for research and analytical applications. It acts as a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant also functions as a GPR30 agonist, effectively inhibiting the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM, inducing autophagy, and demonstrating antitumor efficacy.
  • Analytical standard for research and analytical applications
  • Potent estrogen receptor (ER) antagonist
  • GPR30 agonist
  • Inhibits growth of ER-positive MCF-7 cells
  • Induces autophagy and demonstrates antitumor efficacy
  • Solid appearance
  • White to off-white color
  • Used in HPLC, GC, and MS research experiments