Learn More
Medchemexpress LLC Fulvestrant (standard) | 129453-61-8 | MFCD00903953 | 99.7% | C32H47F5O3S | 10MG
SDP
Catalog No. 5000261169
Encompass_Preferred
Fulvestrant (Standard) is an analytical reference standard of the estrogen receptor antagonist fulvestrant supplied as a 10 mg vial for laboratory analytical use. It is intended for calibration, method development, and quality control in HPLC, LC-MS, and related assays, and is accompanied by supporting documentation for reference use.
  • High purity (>99.7%).
  • Suitable for HPLC and LC-MS method development.
  • Provided as a 10 mg reference vial for analytical workflows.
  • Accompanied by SDS and certificate of analysis.
  • Intended for analytical and quality control use only.
Catalog No. 50-002-61169 Supplier Medchemexpress LLC Supplier No. HY13636R10MG
Fulvestrant (Standard) is an analytical reference standard of the estrogen receptor antagonist fulvestrant supplied as a 10 mg vial for laboratory analytical use. It is intended for calibration, method development, and quality control in HPLC, LC-MS, and related assays, and is accompanied by supporting documentation for reference use.
  • High purity (>99.7%).
  • Suitable for HPLC and LC-MS method development.
  • Provided as a 10 mg reference vial for analytical workflows.
  • Accompanied by SDS and certificate of analysis.
  • Intended for analytical and quality control use only.