Learn More
Cayman Chemical Fulvestrnt 9-sulfon 10mg
SDP
Catalog No. 502912396
Encompass_Preferred
An estrogen receptor antagonist and a derivative of fulvestrant; binds to ERα and ERβ and inhibits estrogen-induced gene expression in a reporter assay from 0.1-100 µM
Catalog No. 50-291-2396 Supplier Cayman Chemical Supplier No. 3507010 MG
An estrogen receptor antagonist and a derivative of fulvestrant; binds to ERα and ERβ and inhibits estrogen-induced gene expression in a reporter assay from 0.1-100 µM