Learn More
AdipoGen Fulvic acid
SDP
Supplier:  AdipoGen AGCN20135M001
Encompass_Preferred
Chemical. CAS 479-66-3. Formula C14H12O8. MW 308.2. Isolated from Penicillium sp. strain FKP-0046. Free radical scavenger and antioxidant. Potential compound to neutralize radioactive and toxic wastes and "heal" soils. Chelates and binds scores of minerals into a bioavailable form used by cells. Shown to incorporate into bone and cartilage of rats. Antibacterial. Antifungal. beta-Hexosaminidase release inhibitor. Anti-inflammatory. Tau fibrils aggregation inhibitor Alzheimers Disease. Antiseptic. Interrupts the dimer formation of Abeta17-42 peptides.
Catalog No. 50-256-2653