Learn More
Abcam Fulvic acid, colloidal poly-electrolyte, 1MG
Catalog No. 502705943
Encompass_Preferred
MW 308.24 Da, Purity >95%. Cell-permeable colloidal poly-electrolyte. Can act as an electron donor or acceptor / oxidizer or reducer. Inhibits aggregation and promotes disassembly of tau fibrils. Shows broad-spectrum antibacterial activity in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5943 Supplier Abcam Supplier No. AB1442561MG
MW 308.24 Da, Purity >95%. Cell-permeable colloidal poly-electrolyte. Can act as an electron donor or acceptor / oxidizer or reducer. Inhibits aggregation and promotes disassembly of tau fibrils. Shows broad-spectrum antibacterial activity in vivo. .
The product is subject to the following: Abcam Restricted Use Statement