Learn More
Medchemexpress LLC Fumagilin-105 | 2410081-58-0 | >97.34% | 784.98 | 1 MG
SDP
Catalog No. 5000393163
Encompass_Preferred
Fumagilin-105 is an autophagy-targeting chimera (AUTOTAC) that degrades MetAP2 via p62-mediated macroautophagy in a ubiquitination-independent manner. It can inhibit the migration of tumor cells and induce programmed cell death, demonstrating anti-tumor activity.
  • Autophagy-targeting chimera (AUTOTAC)
  • Degrades MetAP2 via p62-mediated macroautophagy
  • Utilizes a ubiquitination-independent degradation mechanism
  • Inhibits the migration of tumor cells
  • Induces programmed cell death
  • Possesses anti-tumor activity
Catalog No. 50-003-93163 Supplier Medchemexpress LLC Supplier No. HY1617421MG
Fumagilin-105 is an autophagy-targeting chimera (AUTOTAC) that degrades MetAP2 via p62-mediated macroautophagy in a ubiquitination-independent manner. It can inhibit the migration of tumor cells and induce programmed cell death, demonstrating anti-tumor activity.
  • Autophagy-targeting chimera (AUTOTAC)
  • Degrades MetAP2 via p62-mediated macroautophagy
  • Utilizes a ubiquitination-independent degradation mechanism
  • Inhibits the migration of tumor cells
  • Induces programmed cell death
  • Possesses anti-tumor activity