Learn More
Selleck Chemical LLC Fumagillin 5mg 23110-15-8
SDP
Supplier:  Selleck Chemical LLC S77845MG
Encompass_Preferred
Fumagillin is a selective and potent irreversible inhibitor of Methionine aminopeptidase 2 (MetAP2), used as an antibiotic to treat microsporidiosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5247