Learn More
Enzo Life Sciences Fumagillin (5mg). CAS: 23110-15-8
SDP
Supplier:  Enzo Life Sciences BMLCT1000005
Encompass_Preferred
Inhibitor of angiogenesis and endothelial cell proliferation. Specific inhibitor of methionine aminopeptidase type II (MetAP-II) by covalently modifying a conserved active-site histidine. Precursor to TNP-470. Exhibits antitumor activity in estrogen-induced pituitary adenoma. Antineoplastic. Anti-infective. Alternative name: Fumidil B, Fumadil B, TNP-470. Purity: ≥95% (TLC). Solubility: Soluble in DMSO (25mg/ml) or diluted aqueous bases; slightly soluble in methanol or other alcoholic solvents; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0478