Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumaramide, 98%, Thermo Scientific™

Catalog No. AC338300250
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC338300250 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC338300250 Supplier Thermo Scientific Chemicals Supplier No. 338300250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 627-64-5
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H6N2O2
Linear Formula HNC(O)CHCHC(O)nH
Quantity 25g
MDL Number MFCD00025527
Synonym fumaramide, fumaric diamide, 2-butenediamide, e, 2-butenediamide e, but-2-enediamide, e-but-2-enediamide, 2-butenediamide, 2e, 1e ethene-1,2-dicarboxamide, fumaramid, e-2-butenediamide
InChI Key BSSNZUFKXJJCBG-OWOJBTEDSA-N
SMILES C(=CC(=O)N)C(=O)N
IUPAC Name (E)-but-2-enediamide
Molecular Weight (g/mol) 114.104
PubChem CID 446408
Formula Weight 114.1
Percent Purity ≥97.5%
Chemical Name or Material Fumaramide, 98%
Show More Show Less
EINECSNumber 211-007-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.