Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumaramide 96.0+%, TCI America™
SDP

Catalog No. F006625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F006625G 25 g
F006610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F006625G Supplier TCI America Supplier No. F006625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 627-64-5
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.104
MDL Number MFCD00025527
InChI Key BSSNZUFKXJJCBG-OWOJBTEDSA-N
Synonym fumaramide, fumaric diamide, 2-butenediamide, e, 2-butenediamide e, but-2-enediamide, e-but-2-enediamide, 2-butenediamide, 2e, 1e ethene-1,2-dicarboxamide, fumaramid, e-2-butenediamide
PubChem CID 446408
IUPAC Name (E)-but-2-enediamide
SMILES C(=CC(=O)N)C(=O)N

Specifications

Color White
Quantity 25 g
Formula Weight 114.10
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Fumaramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.