Learn More
Medchemexpress LLC Fumarate hydratase-I 10mg
SDP
Catalog No. 5000168925
Encompass_Preferred
Fumarate hydratase-IN-1 (compound 2) is a cell-permeable fumarate hydratase inhibitor Fumarate hydratase-IN-1 has antiproliferative activity against several cancer cell lines with a mean IC50 of 2 2 M1]
Catalog No. 50-001-68925 Supplier Medchemexpress LLC Supplier No. HY10000410MG
Fumarate hydratase-IN-1 (compound 2) is a cell-permeable fumarate hydratase inhibitor Fumarate hydratase-IN-1 has antiproliferative activity against several cancer cell lines with a mean IC50 of 2 2 M1]