Learn More
TARGETMOL CHEMICALS INC Fumarate hydratase-IN-1 2MG
SDP
Catalog No. 502877589
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Fumarate hydratase-IN-1 is a cell-permeable fumarate hydratase inhibitor with antiproliferative activity and can be used to study cellular activity. Purity 98.96%
Catalog No. 50-287-7589 Supplier TARGETMOL CHEMICALS INC Supplier No. T113372MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Fumarate hydratase-IN-1 is a cell-permeable fumarate hydratase inhibitor with antiproliferative activity and can be used to study cellular activity. Purity 98.96%