Learn More
Medchemexpress LLC HY-100004 5mg , Fumarate hydratase-IN-1 CAS:1644060-37-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1000045MG
Encompass_Preferred
HY-100004 5mg Fumarate hydratase-IN-1 CAS: 1644060-37-6 Fumarate hydratase-IN-1 (compound 2) is a cell-permeable fumarate hydratase inhibitor. Fumarate hydratase-IN-1 has antiproliferative activity against several cancer cell lines with a mean IC50 of 2.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0341