Learn More
Selleck Chemical LLC Fumarate hydratase-IN-1
SDP
Supplier:  Selleck Chemical LLC E1403100MG
Encompass_Preferred
Fumarate hydratase-IN-1 is a small molecule cell-permeable inhibitor of fumarate hydratase It induce alteration of TCA cycle metabolites modulation of cytosolic processes and substantial redox stress responses
Catalog No. 50-313-7724