Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fumarate hydratase-IN-2 (sodium salt) | 2070009-45-7 | 98.7% | 440.47 | C25H25N2NaO4 | 100 MG
SDP

Catalog No. 5000215573
Encompass_Preferred

Fumarate hydratase-IN-2 (sodium salt) is a cell-permeable, competitive inhibitor of fumarate hydratase with a reported Ki of 4.5 μM and nutrient-dependent cytotoxicity. Supplied as a sodium salt, it is intended for biochemical and cell-based studies probing fumarate hydratase activity and related metabolic pathways.

  • Cell-permeable, competitive inhibitor of fumarate hydratase (Ki = 4.5 μM).
  • Reported nutrient-dependent cytotoxicity in cell assays.
  • High purity suitable for research use (LCMS 98.7%).
  • Molecular weight 440.47; formula C25H25N2NaO4.
  • Available in small-scale solid quantities for assay preparation (for example, 100 mg).

Catalog No. 50-002-15573 Supplier Medchemexpress LLC Supplier No. HY100005A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fumarate hydratase-IN-2 (sodium salt) is a cell-permeable, competitive inhibitor of fumarate hydratase with a reported Ki of 4.5 μM and nutrient-dependent cytotoxicity. Supplied as a sodium salt, it is intended for biochemical and cell-based studies probing fumarate hydratase activity and related metabolic pathways.

  • Cell-permeable, competitive inhibitor of fumarate hydratase (Ki = 4.5 μM).
  • Reported nutrient-dependent cytotoxicity in cell assays.
  • High purity suitable for research use (LCMS 98.7%).
  • Molecular weight 440.47; formula C25H25N2NaO4.
  • Available in small-scale solid quantities for assay preparation (for example, 100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.