Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fumarate hydratase-IN-2 sodium salt | 2070009-45-7 | 98.7% | 440.47 g/mol | C25H25N2NaO4 | 1 ML
SDP

Catalog No. 5000215664
Encompass_Preferred

Fumarate hydratase-IN-2 sodium salt is a cell-permeable, competitive inhibitor of fumarate hydratase (Ki = 4.5 μM) used for biochemical and cell-based research into enzyme function and metabolic responses. It is supplied in high-purity form with solution and solid formats to support varied experimental workflows.

  • Cell-permeable competitive inhibitor of fumarate hydratase (Ki = 4.5 μM).
  • Reported nutrient-dependent cytotoxicity in cell studies.
  • High purity suitable for research applications (98.7% reported).
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities for flexibility.
  • Molecular formula C25H25N2NaO4 and molecular weight 440.47 g/mol.
  • Includes product data sheet and certificate of analysis for batch-specific quality assurance.

Catalog No. 50-002-15664 Supplier Medchemexpress LLC Supplier No. HY100005A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fumarate hydratase-IN-2 sodium salt is a cell-permeable, competitive inhibitor of fumarate hydratase (Ki = 4.5 μM) used for biochemical and cell-based research into enzyme function and metabolic responses. It is supplied in high-purity form with solution and solid formats to support varied experimental workflows.

  • Cell-permeable competitive inhibitor of fumarate hydratase (Ki = 4.5 μM).
  • Reported nutrient-dependent cytotoxicity in cell studies.
  • High purity suitable for research applications (98.7% reported).
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities for flexibility.
  • Molecular formula C25H25N2NaO4 and molecular weight 440.47 g/mol.
  • Includes product data sheet and certificate of analysis for batch-specific quality assurance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.