Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fumarate hydratase-IN-2 sodium salt | 2070009-45-7 | MFCD00218071 | 98.7% | 440.47 g/mol | C25H25N2NaO4 | 50 MG
SDP

Catalog No. 5000215130
Encompass_Preferred

Fumarate hydratase-IN-2 sodium salt is a cell-permeable, competitive inhibitor of fumarate hydratase with a reported Ki of 4.5 μM. Supplied as a sodium salt solid, it is intended for biochemical and cellular studies probing fumarate hydratase activity and nutrient-dependent cytotoxicity.

  • Cell-permeable competitive inhibitor of fumarate hydratase.
  • Reported Ki 4.5 μM for fumarate hydratase.
  • Molecular formula C25H25N2NaO4; molecular weight 440.47 g/mol.
  • High purity (98.7%) in solid form.
  • Available in multiple sizes, including 50 mg and 10 mM/1 mL solution.

Catalog No. 50-002-15130 Supplier Medchemexpress LLC Supplier No. HY100005A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fumarate hydratase-IN-2 sodium salt is a cell-permeable, competitive inhibitor of fumarate hydratase with a reported Ki of 4.5 μM. Supplied as a sodium salt solid, it is intended for biochemical and cellular studies probing fumarate hydratase activity and nutrient-dependent cytotoxicity.

  • Cell-permeable competitive inhibitor of fumarate hydratase.
  • Reported Ki 4.5 μM for fumarate hydratase.
  • Molecular formula C25H25N2NaO4; molecular weight 440.47 g/mol.
  • High purity (98.7%) in solid form.
  • Available in multiple sizes, including 50 mg and 10 mM/1 mL solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.