Learn More
Medchemexpress LLC Fumaric acid | 110-17-8 | MFCD00002700 | 99.9% | 116.07 | C4H4O4 | 100 MG
SDP
Catalog No. 5000303884
Encompass_Preferred
Fumaric acid (Standard) is an analytical reference material of fumaric acid (CAS 110-17-8) intended for research and analytical applications. It is supplied as a white to off-white solid used in biochemical studies and as a reference compound for analytical methods.
  • Analytical standard for method development and quality control.
  • High purity suitable for analytical and research use.
  • White to off-white solid, easy to handle and store.
  • Available in multiple pack sizes, including 100 mg, for laboratory convenience.
  • Suitable as a biochemical reference and assay calibrant.
Catalog No. 50-003-03884 Supplier Medchemexpress LLC Supplier No. HYW015883R100MG
Fumaric acid (Standard) is an analytical reference material of fumaric acid (CAS 110-17-8) intended for research and analytical applications. It is supplied as a white to off-white solid used in biochemical studies and as a reference compound for analytical methods.
  • Analytical standard for method development and quality control.
  • High purity suitable for analytical and research use.
  • White to off-white solid, easy to handle and store.
  • Available in multiple pack sizes, including 100 mg, for laboratory convenience.
  • Suitable as a biochemical reference and assay calibrant.