Learn More
Medchemexpress LLC Fumaric acid | 110-17-8 | MFCD00002700 | 99.9% | 116.07 g/mol | C4H4O4 | 50 G
SDP
Catalog No. 5000300263
Encompass_Preferred
Fumaric acid is an unsaturated dicarboxylic acid (trans-butenedioic acid, CAS 110-17-8) used as a reagent in biochemical and pharmacological research. It is typically supplied as a white solid with high reported purity suitable for laboratory applications and studies of metabolic and inflammatory pathways.
  • High purity (99.9%).
  • White crystalline solid suitable for reagent use.
  • Soluble in water and DMSO.
  • Used in metabolic and anti-inflammatory research applications.
  • Available in multiple package sizes, including 50 g.
Catalog No. 50-003-00263 Supplier Medchemexpress LLC Supplier No. HYW01588350G
Fumaric acid is an unsaturated dicarboxylic acid (trans-butenedioic acid, CAS 110-17-8) used as a reagent in biochemical and pharmacological research. It is typically supplied as a white solid with high reported purity suitable for laboratory applications and studies of metabolic and inflammatory pathways.
  • High purity (99.9%).
  • White crystalline solid suitable for reagent use.
  • Soluble in water and DMSO.
  • Used in metabolic and anti-inflammatory research applications.
  • Available in multiple package sizes, including 50 g.