Learn More
Medchemexpress LLC Fumaric acid-13C2 | 96503-56-9 | 99.9% | 118.06 g·mol-1 | C4H4O4 (13C2) | 1 MG
SDP
Catalog No. 5000300982
Encompass_Preferred
Fumaric acid-13C2 is a stable isotope-labeled form of fumaric acid containing two 13C atoms, provided as a white to off-white solid for use as an analytical standard and tracer in mass-spectrometry and metabolic studies. The material is characterized and documented to support reproducible analytical results.
  • Stable isotope labeled with two 13C atoms.
  • High purity (99.9%).
  • White to off-white solid, easy to handle and weigh.
  • Suitable as an internal standard for LC-MS and GC-MS assays.
  • Useful for metabolic flux and tracer experiments.
  • Available in small milligram pack sizes for analytical use.
  • Manufacturer documentation includes SDS and COA for quality assurance.
Catalog No. 50-003-00982 Supplier Medchemexpress LLC Supplier No. HYW015883S11MG
Fumaric acid-13C2 is a stable isotope-labeled form of fumaric acid containing two 13C atoms, provided as a white to off-white solid for use as an analytical standard and tracer in mass-spectrometry and metabolic studies. The material is characterized and documented to support reproducible analytical results.
  • Stable isotope labeled with two 13C atoms.
  • High purity (99.9%).
  • White to off-white solid, easy to handle and weigh.
  • Suitable as an internal standard for LC-MS and GC-MS assays.
  • Useful for metabolic flux and tracer experiments.
  • Available in small milligram pack sizes for analytical use.
  • Manufacturer documentation includes SDS and COA for quality assurance.