Learn More
Medchemexpress LLC Fumaric acid-13C2 | 96503-56-9 | 99.9% | 118.06 g/mol | C2(13C)2H4O4 | 5 MG
SDP
Catalog No. 5000299621
Encompass_Preferred
Fumaric acid-13C2 (CAS 96503-56-9) is the 13C-labeled form of fumaric acid used as an isotopic tracer and internal standard in analytical and biochemical research. With high isotopic enrichment, it supports accurate quantitation and metabolic pathway studies, including citric acid cycle analysis and investigations of NF-κB-dependent signaling.
  • Isotopic tracer for quantitative NMR, GC-MS, and LC-MS analysis.
  • High isotopic enrichment (99.9%) for improved accuracy.
  • Suitable for metabolic and signaling pathway research.
  • Provided in research-scale quantities for analytical workflows.
Catalog No. 50-002-99621 Supplier Medchemexpress LLC Supplier No. HYW015883S15MG
Fumaric acid-13C2 (CAS 96503-56-9) is the 13C-labeled form of fumaric acid used as an isotopic tracer and internal standard in analytical and biochemical research. With high isotopic enrichment, it supports accurate quantitation and metabolic pathway studies, including citric acid cycle analysis and investigations of NF-κB-dependent signaling.
  • Isotopic tracer for quantitative NMR, GC-MS, and LC-MS analysis.
  • High isotopic enrichment (99.9%) for improved accuracy.
  • Suitable for metabolic and signaling pathway research.
  • Provided in research-scale quantities for analytical workflows.