Learn More
Medchemexpress LLC Fumaric acid-13C2,d2 | 100858-52-4 | 98.0% | 120.07 g/mol | C213C2H2D2O4 | 1 MG
SDP
Catalog No. 5000300555
Encompass_Preferred
Fumaric acid-13C2,d2 is a stable isotope-labeled fumaric acid where two carbon atoms are 13C-labeled and two hydrogen atoms are deuterated. It is provided as a high-purity solid for use as an analytical standard, tracer, or reagent in metabolic and mass spectrometry studies, and is available in small milligram quantities for sensitive experiments.
  • Stable isotope labeling for tracer studies and metabolic experiments.
  • High purity suitable for analytical and mass spectrometry applications.
  • Available in small quantities for sensitive assays and method development.
  • Solid, white to off-white appearance for straightforward handling and storage.
  • Provided with product datasheet and safety data sheet for laboratory use.
Catalog No. 50-003-00555 Supplier Medchemexpress LLC Supplier No. HYW015883S21MG
Fumaric acid-13C2,d2 is a stable isotope-labeled fumaric acid where two carbon atoms are 13C-labeled and two hydrogen atoms are deuterated. It is provided as a high-purity solid for use as an analytical standard, tracer, or reagent in metabolic and mass spectrometry studies, and is available in small milligram quantities for sensitive experiments.
  • Stable isotope labeling for tracer studies and metabolic experiments.
  • High purity suitable for analytical and mass spectrometry applications.
  • Available in small quantities for sensitive assays and method development.
  • Solid, white to off-white appearance for straightforward handling and storage.
  • Provided with product datasheet and safety data sheet for laboratory use.